ROX Dye qPCR Calibration Solution *10000X*
Lieferant: AAT BIOQUEST
67034
AATB67034EA
420
EUR
AATB67034
ROX Dye qPCR Calibration Solution *10000X*
Nukleinsäure-Reagenzien
qPCR/RT-qPCR Enzyme und Kits
ROX dye qPCR Calibration Solution supplied as a 10000X concentrate is compatible with a wide variety of qPCR instruments, including the ABI7500 Fast, QuantStudio™, and ViiA™ 7 systems.
- Stable for 12 months
Real-time thermal cyclers require calibration to generate a spectral profile for the fluorescence signal.
Bitte beachten For research use only.