Rhodamin B ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-yliden]diethylazaniumchlorid, 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthyliumchlorid

69
AATB69EA 120 EUR
AATB69
Rhodamin B ≥95%
Rhodamin B

Rhodamine B (also called Rhodamine 610, Basic Violet 10, or C.I.

Formel: C₂₈H₃₁ClN₂O₃
Molecular Weight: 479,02 g/mol
MDL: MFCD00011931
CAS-Nummer: 81-88-9
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR