AML Cy5 NHS ester, ReadiCleave™ zur reversiblen Fluoreszenzmarkierung
Lieferant: AAT BIOQUEST
|
Achtung
|
7000
AATB7000EA
614
EUR
AATB7000
AML Cy5 NHS ester, ReadiCleave™ zur reversiblen Fluoreszenzmarkierung
AML Cy5 NHS ester
Fluorescence-based methods have many advantages for biological detections in terms of sensitivity.
- Reversible linker
- Stable signal
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...