XFD532 AML-NHS ester, ReadiCleave™
Lieferant: AAT Bioquest
ReadiCleave™ AML XFD532 contains an azidomethyl linker that can be cleaved with TCEP to remove the XFD532 fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only