iFluor 700 AML-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7003
AATB7003EA
574
EUR
AATB7003
iFluor 700 AML-NHS ester, ReadiCleave™
iFluor 700 AML-NHS ester
ReadiCleave™ AML iFluor® 700 contains an azidomethyl linker that can be cleaved with TCEP to remove the iFluor® 700 fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only