iFluor 594 AML-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7007
AATB7007EA
574
EUR
AATB7007
iFluor 594 AML-NHS ester, ReadiCleave™
iFluor 594 AML-NHS ester
ReadiCleave™ iFluor® 594 AML uses an azidomethyl linker that can be cleaved with TCEP to remove the iFluor® 594 fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only