XFD488 AML-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7008
AATB7008EA
561
EUR
AATB7008
XFD488 AML-NHS ester, ReadiCleave™
XFD488 AML-NHS ester
ReadiCleave™ AML XFD488 contains an azidomethyl linker that can be cleaved with TCEP to remove the Alexa Fluor® 488 fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only