XFD546 AML-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7009
AATB7009EA
561
EUR
AATB7009
XFD546 AML-NHS ester, ReadiCleave™
XFD546 AML-NHS ester
ReadiCleave™ AML XFD546 contains an azidomethyl linker that can be cleaved with TCEP to remove the Alexa Fluor® 546 fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only