3-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)propionic acid NHS ester ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: BDP FL NHS ester, 3-[3-[(2,5-Dioxopyrrolidin-1-yl)oxy]-3-oxopropyl]-5,5-difluoro-7,9-dimethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide, 7-(3-((2,5-Dioxopyrrolidin-1-yl)oxy)-3-oxopropyl)-5,5-difluoro-1,3-dimethyl-5H-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-4-ium-5-uide, 4,4-Difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-propionic acid N-Succinimidyl ester, Bodi Fluor™ 488 SE

701
AATB701EA 307 EUR
AATB701
3-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)propionic acid NHS ester ≥90%
3-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)propionic acid NHS ester

Bodi Fluor™ 488, SE is the same molecule to Bodipy® FL, SE (Bodipy® is the trademark of Invitrogen).

Formel: C₁₈H₁₈BF₂N₃O₄
Molecular Weight: 389,166 g/mol
MDL: MFCD00467385
CAS-Nummer: 146616-66-2
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR