XFD647 AML-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7012
AATB7012EA 574 EUR
AATB7012
XFD647 AML-NHS ester, ReadiCleave™
XFD647 AML-NHS ester
ReadiCleave™ AML XFD647 contains an azidomethyl linker that can be cleaved with TCEP to remove the Alexa Fluor® 647 fluorophore from the conjugated molecule.
  • Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only