iFluor 488 SSL-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7040
AATB7040EA 561 EUR
AATB7040
iFluor 488 SSL-NHS ester, ReadiCleave™
iFluor 488 SSL-NHS ester
ReadiCleave™ iFluor™ 488 SSL-NHS ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the iFluor™ 488 fluorophore from the target molecule.
  • Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only