FAM SSL-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7041
AATB7041EA
561
EUR
AATB7041
FAM SSL-NHS ester, ReadiCleave™
FAM SSL-NHS ester
ReadiCleave™ FAM SSL-NHS ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the fluorescein (FITC) fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only