Cy5-SSL-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7050
AATB7050EA
561
EUR
AATB7050
Cy5-SSL-NHS ester, ReadiCleave™
Cy5-SSL-NHS ester
ReadiCleave™ Cy5-SSL-NHS ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the Cy5 fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only