XFD532 SSL-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7051
AATB7051EA
490
EUR
AATB7051
XFD532 SSL-NHS ester, ReadiCleave™
XFD532 SSL-NHS ester
ReadiCleave™ XFD 532 succinimidyl ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the Alexa Fluor 532 fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only