ROXtra SSL-NHS ester, ReadiCleave™
Lieferant: AAT BIOQUEST
7052
AATB7052EA
489
EUR
AATB7052
ROXtra SSL-NHS ester, ReadiCleave™
ROXtra SSL-NHS ester
ReadiCleave™ ROXtra succinimidyl ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the ROXtra fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Bitte beachten For Research Use Only