5-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 5-isomer]

Lieferant: AAT BIOQUEST

Synonyme: Oregon Green 488 carboxylic acid, 5-isomer

708
AATB708EA 307 EUR
AATB708
5-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 5-isomer]
5-OG488 acid

5-OG488 acid is the same molecule to Oregon Green® 488 carboxylic acid, 5-isomer (Oregon Green® is the Trademark of Invitrogen).


  • Photostable
  • Bright Fluorescence
  • pH-independent Fluorescence
  • 95% Purity


Bitte beachten For research use only.

CAS-Nummer: 1027168-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR