Sulphorhodamine 101 sodium salt ≥95%
Lieferant: AAT BIOQUEST
71
AATB71EA
120
EUR
AATB71
Sulphorhodamine 101 sodium salt ≥95%
Sulphorhodamine 101 sodium salt
Sulforhodamine 101 is a fluorescence reference standard for measuring fluorescence quantum yield.
- Fluorescence Reference Standard
- Water Soluble
Bitte beachten For research use only.
CAS-Nummer:
465544-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...