iFluor® 633, Succinimidyl Ester
Lieferant: AAT Bioquest
1030
71030
71509
71559
AATB1030EA
307
EUR
AATB1030
AATB71030
AATB71509
AATB71559
iFluor® 633, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- pH-Independent Signal
Bitte beachten For research use only.