iFluor® 633, Succinimidyl Ester
Lieferant: AAT Bioquest
1030 71030 71509 71559
AATB1030EA 307 EUR
AATB1030 AATB71030 AATB71509 AATB71559
iFluor® 633, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Bitte beachten For research use only.