iFluor® 700, Succinimidyl Ester
Lieferant: AAT BIOQUEST
1036 71036 71514 71564
AATB1036EA 307 EUR
AATB1036 AATB71036 AATB71514 AATB71564
iFluor® 700, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Bitte beachten For research use only.