iFluor® 700, Succinimidyl Ester
Lieferant: AAT BIOQUEST
1036
71036
71514
71564
AATB1036EA
307
EUR
AATB1036
AATB71036
AATB71514
AATB71564
iFluor® 700, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- pH-Independent Signal
Bitte beachten For research use only.