6-OG488 succinimidyl ester ≥90% [equivalent to Oregon Green® 488 carboxylic acid, succinimidyl ester, 6-isomer ]
Lieferant: AAT BIOQUEST
Synonyme:
Oregon Green 488 carboxylic acid succinimidyl ester, 6-isomer
711
AATB711EA
307
EUR
AATB711
6-OG488 succinimidyl ester ≥90% [equivalent to Oregon Green® 488 carboxylic acid, succinimidyl ester, 6-isomer ]
6-OG488 succinimidyl ester
6-OG488 succinimidyl ester is the same molecule to Oregon Green® 488 carboxylic acid, succinimidyl ester, 6-isomer (Invitrogen #O6185, Oregon Green® is the Trademark of Invitrogen).
- Photostable
- Bright Fluorescence
- pH-independent Fluorescence
- 90% Purity
Bitte beachten For research use only.
CAS-Nummer:
198139-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...