6-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 6-isomer]

Lieferant: AAT BIOQUEST

Synonyme: Oregon Green 488 carboxylic acid, 6-isomer

712
AATB712EA 307 EUR
AATB712
6-OG488 acid ≥95% [equivalent to Oregon Green® 488 carboxylic acid, 6-isomer]
6-OG488 acid

6-OG488 acid is the same molecule to Oregon Green® 488 carboxylic acid, 6-isomer (Oregon Green® is the Trademark of Invitrogen).


  • Photostable
  • Bright Fluorescence
  • pH-independent Fluorescence
  • 95% Purity


Bitte beachten For research use only.

CAS-Nummer: 195136-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR