OG488 maleimide ≥90% [equivalent to Oregon Green® 488 maleimide]

Lieferant: AAT BIOQUEST

Synonyme: Oregon Green 488 maleimide

715
AATB715EA 307 EUR
AATB715
OG488 maleimide ≥90% [equivalent to Oregon Green® 488 maleimide]
OG488 maleimide

OG488 maleimide is the same molecule to Oregon Green® 488 maleimide (Invitrogen #O6034, Oregon Green® is the Trademark of Invitrogen).


  • Photostable
  • Bright Fluorescence
  • pH-independent Fluorescence
  • 90% Purity


Bitte beachten For research use only.

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR