iFluor® 820, Succinimidyl Ester
Lieferant: AAT Bioquest
1399 71399 71527 71577
AATB1399EA 460 EUR
AATB1399 AATB71399 AATB71527 AATB71577
iFluor® 820, Succinimidyl Ester
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • Useful For In-Vivo Imaging
Bitte beachten For research use only.