iFluor® 594, Succinimidyl Ester
Lieferant: AAT BIOQUEST
71029 1029 71508 71558
AATB71029EA 114 EUR
AATB71029 AATB1029 AATB71508 AATB71558
iFluor® 594, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Bitte beachten For research use only.