iFluor® 594, Succinimidyl Ester
Lieferant: AAT BIOQUEST
71029
1029
71508
71558
AATB71029EA
114
EUR
AATB71029
AATB1029
AATB71508
AATB71558
iFluor® 594, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright Fluorescence
- High Photostability
- Low Quenching On Proteins
- pH-Independent Signal
Bitte beachten For research use only.