iFluor® Ultra 594 maleimide
Lieferant: AAT BIOQUEST
71653
AATB71653EA 470 EUR
AATB71653
iFluor® Ultra 594 maleimide
iFluor® Ultra 594 maleimide
iFluor® Ultra 594 is an exceptionally bright red fluorescent dye excitable by 561 nm or 594 nm laser lines, with a peak emission at 601 nm.
  • Stable for 12 months
The iFluor® Ultra series represents an enhancement of our established iFluor® dyes, optimized for antibody labeling in fluorescence imaging and flow cytometry.
Bitte beachten For Research Use Only