iFluor® Ultra 647 maleimide
Lieferant: AAT BIOQUEST
71673
AATB71673EA
470
EUR
AATB71673
iFluor® Ultra 647 maleimide
iFluor® Ultra 647 maleimide
iFluor® Ultra 647 is an exceptionally bright far-red fluorescent dye excitable by 594 nm or 633 nm laser lines, with a peak emission at 670 nm.
- Stable for 12 months
The iFluor® Ultra series represents an enhancement of our established iFluor® dyes, optimized for antibody labeling in fluorescence imaging and flow cytometry.
Bitte beachten For Research Use Only