OG514 succinimidyl ester [equivalent to Oregon Green® 514 succinimidyl ester]
Lieferant: AAT Bioquest
717
AATB717EA
401
EUR
AATB717
OG514 succinimidyl ester [equivalent to Oregon Green® 514 succinimidyl ester]
OG514 succinimidyl ester
OG514 is the same fluorophore as Oregon Green 514 (Oregon Green® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only