XFD555 NHS ester [Same Structure to Alexa Fluor™ 555 NHS ester]
Lieferant: AAT BIOQUEST
XFD555 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 555 (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only