XFD568 NHS Ester [Same Structure to Alexa Fluor™ 568 NHS ester]

Lieferant: AAT BIOQUEST

71907
AATB71907EA 1100 EUR
AATB71907
XFD568 NHS Ester [Same Structure to Alexa Fluor™ 568 NHS ester]
XFD568 NHS Ester

XFD568 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 568 (Alexa Fluor® is the trademark of ThermoFisher).


  • Stable for 12 months


Quality tested in-house


Bitte beachten For Research Use Only

Molecular Weight: 791,8 g/mol
CAS-Nummer: 247145-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR