XFD594 NHS Ester [Same Structure to Alexa Fluor™ 594 NHS ester]
Lieferant: AAT BIOQUEST
71908
AATB71908EA
1100
EUR
AATB71908
XFD594 NHS Ester [Same Structure to Alexa Fluor™ 594 NHS ester]
XFD594 NHS Ester
XFD594 NHS Ester (Succinimidyl Ester) is the same molecule to the Alexa Fluor® 594 NHS Ester (Alexa Fluor® is the trademark of ThermoFisher).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only