Acid red 52 ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Food Red No.106, Sodium 4-(3,6-bis(diethylamino)xanthylium-9-yl)benzene-1,3-disulphonate, Kiton Red S, Sulphorhodamine B sodium salt, Lissamine rhodamine B-200

72
AATB72EA 120 EUR
AATB72
Acid red 52 ≥95%
Acid red 52

Sulforhodamine B, also known as Kiton Red, is primarily used as a polar tracer.

Formel: C₂₇H₃₀N₂O₇S₂
Molecular Weight: 580,65 g/mol
MDL: MFCD00010180
CAS-Nummer: 3520-42-1
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR