iFluor® 514 maleimide
Lieferant: AAT BIOQUEST
72500
AATB72500EA
309
EUR
AATB72500
iFluor® 514 maleimide
iFluor® 514 maleimide
iFluor® 514 dyes have fluorescence excitation and emission maxima of ~510 nm and ~527 nm respectively.
- Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
Bitte beachten For Research Use Only