iFluor® 660 maleimide
Lieferant: AAT BIOQUEST
72501
AATB72501EA 309 EUR
AATB72501
iFluor® 660 maleimide
iFluor® 660 maleimide
iFluor® 660 dyes have fluorescence excitation and emission maxima of ~660 nm and ~680 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
Bitte beachten For Research Use Only