Cy3 azide, FastClick™
Lieferant: AAT BIOQUEST
72700
AATB72700EA 266 EUR
AATB72700
Cy3 azide, FastClick™
Cy3 azide
FastClick™ Cy3 Azide contains both the moiety of FastClick (for assisting click efficiency) and Cy3 fluorophore (as the fluorescence tag) for developing Cy3-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only