5-FAM azide, FastClick™
Lieferant: AAT BIOQUEST
72710
AATB72710EA 266 EUR
AATB72710
5-FAM azide, FastClick™
5-FAM azide
FastClick™ 5-FAM Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and 5-FAM fluorophore (as the fluorescence tag) for developing 5-FAM-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only