6-FAM Azide, FastClick™
Lieferant: AAT BIOQUEST
72711
AATB72711EA
266
EUR
AATB72711
6-FAM Azide, FastClick™
6-FAM Azide
FastClick™ 6-FAM Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and 6-FAM fluorophore (as the fluorescence tag) for developing 6-FAM-based fluorescent probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only
Molecular Weight:
588.58 g/mol |
CAS-Nummer:
1386385-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...