5-TAMRA azide, FastClick™
Lieferant: AAT Bioquest
FastClick™ 5-TAMRA Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and 5-TAMRA fluorophore (as the fluorescence tag) for developing 5-TAMRA-based fluorescent probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only