6-ROX azide, FastClick™
Lieferant: AAT BIOQUEST
72714
AATB72714EA 266 EUR
AATB72714
6-ROX azide, FastClick™
6-ROX azide
FastClick™ 6-ROX Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and 6-ROX fluorophore (as the fluorescence tag) for developing 6-ROX-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house