XFD350 azide, FastClick™
Lieferant: AAT BIOQUEST
72730
AATB72730EA 266 EUR
AATB72730
XFD350 azide, FastClick™
XFD350 azide
FastClick™ XFD350 Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 350 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 350-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only