XFD405 azide, FastClick™
Lieferant: AAT BIOQUEST
72733
AATB72733EA
266
EUR
AATB72733
XFD405 azide, FastClick™
XFD405 azide
FastClick™ XFD405 Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 405 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 405-based fluorescent probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only