XFD488 azide, FastClick™
Lieferant: AAT BIOQUEST
72735
AATB72735EA
266
EUR
AATB72735
XFD488 azide, FastClick™
XFD488 azide
FastClick™ XFD488 Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 488 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 488-based fluorescent probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only