XFD647 azide, FastClick™
Lieferant: AAT BIOQUEST
72740
AATB72740EA
266
EUR
AATB72740
XFD647 azide, FastClick™
XFD647 azide
FastClick™ XFD647 Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 647 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 647-based fluorescent probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only