Biotin azide, FastClick™
Lieferant: AAT BIOQUEST
72801
AATB72801EA
266
EUR
AATB72801
Biotin azide, FastClick™
Biotin azide
FastClick™ Biotin Azide contains both the CAG moiety of FastClick (for assisting click efficiency) and biotin hapten (as the detection tag) for developing biotin-based probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only