Cy3 alkyne, FastClick™
Lieferant: AAT BIOQUEST
FastClick™ Cy3 Alkyne contains both the moiety of FastClick (for assisting click efficiency) and Cy3 fluorophore (as the fluorescence tag) for developing Cy3-based fluorescent probes.
- Stable for 24 months
Quality tested in-house