5-TAMRA alkyne, FastClick™
Lieferant: AAT BIOQUEST
72862
AATB72862EA
266
EUR
AATB72862
5-TAMRA alkyne, FastClick™
5-TAMRA alkyne
FastClick™ 5-TAMRA Alkyne contains both the moiety of FastClick (for assisting click efficiency) and 5-TAMRA fluorophore (as the fluorescence tag) for developing 5-TAMRA-based fluorescent probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only