6-TAMRA alkyne, FastClick™
Lieferant: AAT BIOQUEST
FastClick™ 6-TAMRA Alkyne contains both the moiety of FastClick (for assisting click efficiency) and 6-TAMRA fluorophore (as the fluorescence tag) for developing 6-TAMRA-based fluorescent probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only
Formel:
C28H25N3O4 |
CAS-Nummer:
1352649-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...