6-ROX alkyne, FastClick™
Lieferant: AAT BIOQUEST
72864
AATB72864EA 266 EUR
AATB72864
6-ROX alkyne, FastClick™
6-ROX alkyne
FastClick™ 6-ROX Alkyne contains both the moiety of FastClick (for assisting click efficiency) and 6-ROX fluorophore (as the fluorescence tag) for developing 6-ROX-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only