XFD350 alkyne, FastClick™
Lieferant: AAT BIOQUEST
72870
AATB72870EA 266 EUR
AATB72870
XFD350 alkyne, FastClick™
XFD350 alkyne
FastClick™ XFD350 Alkyne contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 350 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 350-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only