XFD488 alkyne, FastClick™
Lieferant: AAT BIOQUEST
72875
AATB72875EA 266 EUR
AATB72875
XFD488 alkyne, FastClick™
XFD488 alkyne
FastClick™ XFD488 Alkyne contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 488 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 488-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house