XFD647 alkyne, FastClick™
Lieferant: AAT BIOQUEST
72880
AATB72880EA 266 EUR
AATB72880
XFD647 alkyne, FastClick™
XFD647 alkyne
FastClick™ XFD647 Alkyne contains both the CAG moiety of FastClick (for assisting click efficiency) and Alexa Fluor® 647 fluorophore (as the fluorescence tag) for developing Alexa Fluor® 647-based fluorescent probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only